1,3-Cyclohexanedione 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR28703-100G OR28703-1KG OR28703-500G
APOSOR28703-100GEA 22 EUR
APOSOR28703-100G APOSOR28703-1KG APOSOR28703-500G
1,3-Cyclohexanedione 99%
1,3-Cyclohexanedione
Formule: C₆H₈O₂
Smeltpunt: 101…105 °C
Dichtheid: 1,1 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001585
CAS nummer: 504-02-9
EINECS: 207-980-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR