Formamidine acetate 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR28877-100G OR28877-500G
APOSOR28877-100GEA 58.4 EUR
APOSOR28877-100G APOSOR28877-500G
Formamidine acetate 99%
Formamidine acetate
Formule: C₃H₈N₂O₂
Smeltpunt: 157…161 °C
Dichtheid: 1,124 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012866
CAS nummer: 3473-63-0
EINECS: 222-442-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR