3-Bromophenyl isocyanate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR28901-25G OR28901-5G
APOSOR28901-25GEA 238 EUR
APOSOR28901-25G APOSOR28901-5G
3-Bromophenyl isocyanate 97%
3-Bromophenyl isocyanate

  • May form small amount of sediment.

Formule: C₇H₄BrNO
Kookpunt: 220 °C
Dichtheid: 1,586 g/cm³ (25 °C)
Vlampunt: 108 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002014
CAS nummer: 23138-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR