5-Oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR29064-1G OR29064-5G OR29064-250MG
APOSOR29064-1GEA 131 EUR
APOSOR29064-1G APOSOR29064-5G APOSOR29064-250MG
5-Oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid 95%
5-Oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid
Formule: C₇H₆N₂O₃S
Smeltpunt: 235…238 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00085018
CAS nummer: 32084-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR