Methyl-1H-pyrrole-2-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR29375-100G OR29375-25G
APOSOR29375-100GEA 199 EUR
APOSOR29375-100G APOSOR29375-25G
Methyl-1H-pyrrole-2-carboxylate
Methyl-1H-pyrrole-2-carboxylate
Formule: C₆H₇NO₂
Smeltpunt: 74…78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00817048
CAS nummer: 1193-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR