5-Bromo-1-chloroisoquinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR300068-1G OR300068-250MG
APOSOR300068-1GEA 146 EUR
APOSOR300068-1G APOSOR300068-250MG
5-Bromo-1-chloroisoquinoline
5-Bromo-1-chloroisoquinoline
Formule: C₉H₅BrClN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08703278
CAS nummer: 34551-41-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR