3-Bromo-5-chloroisoquinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR300093-250MG
APOSOR300093-250MGEA 73 EUR
APOSOR300093-250MG
3-Bromo-5-chloroisoquinoline
3-Bromo-5-chloroisoquinoline
Formule: C₉H₅BrClN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18793406
CAS nummer: 1276056-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR