4-Bromoindole-6-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR300307-1G OR300307-250MG
APOSOR300307-1GEA 125 EUR
APOSOR300307-1G APOSOR300307-250MG
4-Bromoindole-6-carboxylic acid
4-Bromoindole-6-carboxylic acid
Formule: C₉H₆BrNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03095205
CAS nummer: 374633-27-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR