Isoxazol-5-ylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR300646-1G OR300646-250MG OR300646-5G
APOSOR300646-1GEA 55.5 EUR
APOSOR300646-1G APOSOR300646-250MG APOSOR300646-5G
Isoxazol-5-ylamine
Isoxazol-5-ylamine
Formule: C₃H₄N₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00128192
CAS nummer: 14678-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR