Ethyl-1-bromocyclobutanecarboxylate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR300697-1G OR300697-25G OR300697-5G
APOSOR300697-1GEA 22 EUR
APOSOR300697-1G APOSOR300697-25G APOSOR300697-5G
Ethyl-1-bromocyclobutanecarboxylate 97%
Ethyl-1-bromocyclobutanecarboxylate
Formule: C₇H₁₁BrO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001322
CAS nummer: 35120-18-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR