Methyl 4-chlorobenzimidate hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR300716-1G
APOSOR300716-1GEA 0 EUR
APOSOR300716-1G
Methyl 4-chlorobenzimidate hydrochloride
Methyl 4-chlorobenzimidate hydrochloride
Formule: C₈H₈ClNO·ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12031863
CAS nummer: 274932-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR