4-Bromo-1,3-thiazole-2-carboxylic acid 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR301250-1G OR301250-5G
APOSOR301250-1GEA 71.6 EUR
APOSOR301250-1G APOSOR301250-5G
4-Bromo-1,3-thiazole-2-carboxylic acid 96%
4-Bromo-1,3-thiazole-2-carboxylic acid
Formule: C₄H₂BrNO₂S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09701413
CAS nummer: 88982-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR