3-Amino-6-bromopyridazine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 6-Bromopyridazin-3-amine

OR302543-25G
APOSOR302543-25GEA 203 EUR
APOSOR302543-25G
3-Amino-6-bromopyridazine 98%
3-Amino-6-bromopyridazine
Formule: C4H4BrN3
MW: 173.9987 g/mol
Smeltpunt: 210 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01646092
CAS nummer: 88497-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR