2,6-Dimethylbenzo[1,2-d:4,5-d']bis[1,3]thiazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR303300-1G OR303300-500MG
APOSOR303300-1GEA 182 EUR
APOSOR303300-1G APOSOR303300-500MG
2,6-Dimethylbenzo[1,2-d:4,5-d']bis[1,3]thiazole
2,6-Dimethylbenzo[1,2-d:4,5-d']bis[1,3]thiazole
Formule: C₁₀H₈N₂S₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07161751
CAS nummer: 13399-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR