4-Iodophenylboronic acid pinacol ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-Iodobenzeneboronic acid pinacol ester, 2-(4-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303432-5G
APOSOR303432-5GEA 114 EUR
APOSOR303432-5G
4-Iodophenylboronic acid pinacol ester
4-Iodophenylboronic acid pinacol ester
Formule: C₁₂H₁₆BIO₂
MW: 329.97 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09953499
CAS nummer: 73852-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR