4-Bromophenylboronic acid pinacol ester 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR303515-5G OR303515-25G
APOSOR303515-5GEA 32.2 EUR
APOSOR303515-5G APOSOR303515-25G
4-Bromophenylboronic acid pinacol ester 97%
4-Bromophenylboronic acid pinacol ester
Formule: C₁₂H₁₆BBrO₂
Smeltpunt: 71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11110553
CAS nummer: 68716-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR