3'-Benzyloxyacetophenone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR30368-25G
APOSOR30368-25GEA 81.8 EUR
APOSOR30368-25G
3'-Benzyloxyacetophenone 98%
3'-Benzyloxyacetophenone
Formule: C₁₅H₁₄O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00026221
CAS nummer: 34068-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR