Cyclohexylacetaldehyde 90%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (2-Oxoethyl)cyclohexane

OR30546-1G OR30546-250MG
APOSOR30546-1GEA 121 EUR
APOSOR30546-1G APOSOR30546-250MG
Cyclohexylacetaldehyde 90%
Cyclohexylacetaldehyde
Formule: C₈H₁₄O
Kookpunt: 58…60 °C (10 mm Hg)
Dichtheid: 0,919 g/cm³ (25 °C)
Vlampunt: 62 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD02261737
CAS nummer: 5664-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR