6-(Bromomethyl)isoquinoline hydrobromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR30570-1G OR30570-100MG OR30570-250MG
APOSOR30570-1GEA 421 EUR
APOSOR30570-1G APOSOR30570-100MG APOSOR30570-250MG
6-(Bromomethyl)isoquinoline hydrobromide
6-(Bromomethyl)isoquinoline hydrobromide
Formule: C₁₀H₈BrN·BrH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12022337
CAS nummer: 188861-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR