2,3-Dimethylhydroquinone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR30633-25G
APOSOR30633-25GEA 151 EUR
APOSOR30633-25G
2,3-Dimethylhydroquinone
2,3-Dimethylhydroquinone
Formule: C₈H₁₀O₂
Smeltpunt: 223 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009997
CAS nummer: 608-43-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR