5-Bromo-2,4-dimethylaniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR30656-1G OR30656-25G OR30656-5G
APOSOR30656-1GEA 29.2 EUR
APOSOR30656-1G APOSOR30656-25G APOSOR30656-5G
5-Bromo-2,4-dimethylaniline
5-Bromo-2,4-dimethylaniline
Formule: C₈H₁₀BrN
Smeltpunt: 96…100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09038481
CAS nummer: 69383-60-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR