3-Bromo-5-azaindole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR30681-1G OR30681-250MG OR30681-5G
APOSOR30681-1GEA 95 EUR
APOSOR30681-1G APOSOR30681-250MG APOSOR30681-5G
3-Bromo-5-azaindole
3-Bromo-5-azaindole
Formule: C₇H₅BrN₂
Smeltpunt: 184…190 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD08690126
CAS nummer: 23612-36-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR