3-Bromo-1H-pyrrolo[2,3-c]pyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR30682-1G OR30682-250MG OR30682-5G
APOSOR30682-1GEA 71.6 EUR
APOSOR30682-1G APOSOR30682-250MG APOSOR30682-5G
3-Bromo-1H-pyrrolo[2,3-c]pyridine
3-Bromo-1H-pyrrolo[2,3-c]pyridine
Formule: C7H5BrN2
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06738910
CAS nummer: 67058-76-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR