1H-Indazole-3-carboxaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR30687-1G OR30687-5G OR30687-10G
APOSOR30687-1GEA 29.2 EUR
APOSOR30687-1G APOSOR30687-5G APOSOR30687-10G
1H-Indazole-3-carboxaldehyde
1H-Indazole-3-carboxaldehyde
Formule: C₈H₆N₂O
Smeltpunt: 141…145 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04972468
CAS nummer: 5235-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR