tert-Butyl 3-bromo-1H-pyrazole-1-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR307167-1G OR307167-5G
APOSOR307167-1GEA 57 EUR
APOSOR307167-1G APOSOR307167-5G
tert-Butyl 3-bromo-1H-pyrazole-1-carboxylate
tert-Butyl 3-bromo-1H-pyrazole-1-carboxylate
Formule: C₈H₁₁BrN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD25976492
CAS nummer: 1448855-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR