1-Chloro-4-cyclopropylbenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307247-1G OR307247-5G
APOSOR307247-1GEA 395 EUR
APOSOR307247-1G APOSOR307247-5G
1-Chloro-4-cyclopropylbenzene
1-Chloro-4-cyclopropylbenzene
Formule: C₉H₉Cl
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD15527177
CAS nummer: 1798-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR