3-Benzo[1,3]dioxol-5-ylmethyl-piperidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307735-1G
APOSOR307735-1GEA 470 EUR
APOSOR307735-1G
3-Benzo[1,3]dioxol-5-ylmethyl-piperidine
3-Benzo[1,3]dioxol-5-ylmethyl-piperidine
Formule: C₁₃H₁₇NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05863607
CAS nummer: 955314-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR