(R)-(-)-Tetrahydrofurfuryl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (R)-Tetrahydro-2-furanmethanol, (R)-(Tetrahydrofuran-2-yl)methanol

OR307767-1G
APOSOR307767-1GEA 106 EUR
APOSOR307767-1G
(R)-(-)-Tetrahydrofurfuryl alcohol
(R)-(-)-Tetrahydrofurfuryl alcohol
Formule: C₅H₁₀O₂
MW: 102.133 g/mol
Kookpunt: 178 °C
Smeltpunt: –80 °C
Dichtheid: 1.054 g/cm³ (25 °C)
Vlampunt: 75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03093085
CAS nummer: 22415-59-4
EINECS: 202-625-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR