4-Amino-8-methoxyquinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR307959-1G OR307959-5G
APOSOR307959-1GEA 328 EUR
APOSOR307959-1G APOSOR307959-5G
4-Amino-8-methoxyquinoline
4-Amino-8-methoxyquinoline
Formule: C₁₀H₁₀N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08063217
CAS nummer: 220844-65-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR