tert-Butyl n-(3-methylpiperidin-3-yl)carbamate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR308123-1G OR308123-5G
APOSOR308123-1GEA 58.4 EUR
APOSOR308123-1G APOSOR308123-5G
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate 97%
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate
Formule: C₁₁H₂₂N₂O₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD13194151
CAS nummer: 169750-96-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR