2-(3-Methyl-5-thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR309408-1G
APOSOR309408-1GEA 478 EUR
APOSOR309408-1G
2-(3-Methyl-5-thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid
2-(3-Methyl-5-thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid
Formule: C₅H₇N₃O₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08437535
CAS nummer: 42047-33-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR