2,7-Dichloroquinoline-3-carbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR309453-1G
APOSOR309453-1GEA 319 EUR
APOSOR309453-1G
2,7-Dichloroquinoline-3-carbonitrile
2,7-Dichloroquinoline-3-carbonitrile
Formule: C₁₀H₄Cl₂N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09787675
CAS nummer: 158583-91-6
UN: 3439
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR