2’-Bromo-4’-iodoacetophenone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR31027-1G OR31027-250MG OR31027-5G
APOSOR31027-1GEA 636 EUR
APOSOR31027-1G APOSOR31027-250MG APOSOR31027-5G
2’-Bromo-4’-iodoacetophenone 95%
2’-Bromo-4’-iodoacetophenone
Formule: C₈H₆BrIO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18391606
CAS nummer: 1261816-56-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR