1-Bromo-2,4-dimethyl-3-nitrobenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR310930-500MG
APOSOR310930-500MGEA 665 EUR
APOSOR310930-500MG
1-Bromo-2,4-dimethyl-3-nitrobenzene
1-Bromo-2,4-dimethyl-3-nitrobenzene
Formule: C₈H₈BrNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00092653
CAS nummer: 39053-43-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR