2-Chloro-4-methyl-5-nitropyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3149-5G
APOSOR3149-5GEA 22 EUR
APOSOR3149-5G
2-Chloro-4-methyl-5-nitropyridine
2-Chloro-4-methyl-5-nitropyridine
Formule: C₆H₅ClN₂O₂
Kookpunt: 91 °C/5mmHg
Smeltpunt: 37…39 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010688
CAS nummer: 23056-33-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR