Pyruvic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR315034-100G OR315034-500G OR315034-2.5KG
APOSOR315034-100GEA 110 EUR
APOSOR315034-100G APOSOR315034-500G APOSOR315034-2.5KG
Pyruvic acid 98%
Pyruvic acid
Formule: C₃H₄O₃
Kookpunt: 164…166 °C
Smeltpunt: 44541 °C
Dichtheid: 1,267 g/cm³ (25 °C)
Vlampunt: 83 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002585
CAS nummer: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR