3-Bromo-p-toluonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3157-25G OR3157-5G OR3157-100G
APOSOR3157-25GEA 24.9 EUR
APOSOR3157-25G APOSOR3157-5G APOSOR3157-100G
3-Bromo-p-toluonitrile 98%
3-Bromo-p-toluonitrile
Formule: C₈H₆BrN
MW: 196.05 g/mol
Smeltpunt: 41…45 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06797818
CAS nummer: 42872-74-2
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR