Methyl 7-amino-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylate 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR318105-1G OR318105-10G OR318105-5G
APOSOR318105-1GEA 506 EUR
APOSOR318105-1G APOSOR318105-10G APOSOR318105-5G
Methyl 7-amino-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylate 95+%
Methyl 7-amino-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylate
Formule: C₁₀H₁₁NO₄
Smeltpunt: 91…93 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03724048
CAS nummer: 20197-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR