2-(Oxiran-2-ylmethoxy)benzonitrile 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR318126-1G OR318126-10G OR318126-5G
APOSOR318126-1GEA 321 EUR
APOSOR318126-1G APOSOR318126-10G APOSOR318126-5G
2-(Oxiran-2-ylmethoxy)benzonitrile 95+%
2-(Oxiran-2-ylmethoxy)benzonitrile
Formule: C₁₀H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01632505
CAS nummer: 38465-16-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR