5-Bromo-1-methyl-1H-indole-3-carbaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR321310-1G OR321310-5G
APOSOR321310-1GEA 125 EUR
APOSOR321310-1G APOSOR321310-5G
5-Bromo-1-methyl-1H-indole-3-carbaldehyde 97%
5-Bromo-1-methyl-1H-indole-3-carbaldehyde
Formule: C₁₀H₈BrNO
Smeltpunt: 133…135 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03906373
CAS nummer: 10102-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR