1,2-Dimethyl (1R,2S)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR32235-1G
APOSOR32235-1GEA 1580 EUR
APOSOR32235-1G
1,2-Dimethyl (1R,2S)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech
1,2-Dimethyl (1R,2S)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate
Formule: C₁₂H₁₈O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02571655
CAS nummer: 55264-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR