(2-Bromopropyl)benzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR323250-25G OR323250-5G
APOSOR323250-25GEA 833 EUR
APOSOR323250-25G APOSOR323250-5G
(2-Bromopropyl)benzene
(2-Bromopropyl)benzene
Formule: C₆H₅CH₂CHBrCH₃
Kookpunt: 107…109 °C
Dichtheid: 1,29 g/cm³ (25 °C)
Vlampunt: 91 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000149
CAS nummer: 2114-39-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR