2-[(4-Chlorophenyl)thio]ethanamine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR32629-1G
APOSOR32629-1GEA 182 EUR
APOSOR32629-1G
2-[(4-Chlorophenyl)thio]ethanamine 95%
2-[(4-Chlorophenyl)thio]ethanamine
Formule: C₈H₁₀ClNS
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00215613
CAS nummer: 36155-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR