2-(Allyloxy)-5-chlorobenzaldehyde 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR32851-1G OR32851-25G OR32851-5G
APOSOR32851-1GEA 182 EUR
APOSOR32851-1G APOSOR32851-25G APOSOR32851-5G
2-(Allyloxy)-5-chlorobenzaldehyde 95%
2-(Allyloxy)-5-chlorobenzaldehyde
Formule: C₁₀H₉ClO₂
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 152842-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR