3-Chlorobenzenecarbaldehyde oxime 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR32889-1G OR32889-5G
APOSOR32889-1GEA 164 EUR
APOSOR32889-1G APOSOR32889-5G
3-Chlorobenzenecarbaldehyde oxime 95%
3-Chlorobenzenecarbaldehyde oxime
Formule: C₇H₆ClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01070635
CAS nummer: 92062-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR