5-Isoquinolineboronic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3296-100MG OR3296-1G
APOSOR3296-100MGEA 32.2 EUR
APOSOR3296-100MG APOSOR3296-1G
5-Isoquinolineboronic acid 97%
5-Isoquinolineboronic acid
Formule: C₉H₈BNO₂
Smeltpunt: 186…188 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03839356
CAS nummer: 371766-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR