6-(1-Pyrrolidinyl)nicotinic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33166-1G OR33166-5G
APOSOR33166-1GEA 338 EUR
APOSOR33166-1G APOSOR33166-5G
6-(1-Pyrrolidinyl)nicotinic acid 95%
6-(1-Pyrrolidinyl)nicotinic acid
Formule: C₁₀H₁₂N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03791234
CAS nummer: 210963-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR