1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33338-1G OR33338-5G
APOSOR33338-1GEA 164 EUR
APOSOR33338-1G APOSOR33338-5G
1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formule: C₁₃H₉Cl₂NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00140302
CAS nummer: 338977-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR