3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR33478-1G OR33478-5G
APOSOR33478-1GEA 338 EUR
APOSOR33478-1G APOSOR33478-5G
3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile 95%
3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile
Formule: C₁₄H₁₇N₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12025842
CAS nummer: 1135282-83-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR