Tetrabutylammonium chloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR33653-1KG OR33653-250G
APOSOR33653-1KGEA 584 EUR
APOSOR33653-1KG APOSOR33653-250G
Tetrabutylammonium chloride 98%
Tetrabutylammonium chloride
Formule: C₁₆H₃₆ClN
MW: 277.92 g/mol
Vlampunt: 110
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011635
CAS nummer: 1112-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR